Gas Chromatography High-Resolution Mass Spectrometer Offers New Opportunities for Analytical Testing
The Thermo Scientific Orbitrap Exploris GC Mass Spectrometer simplifies operations, delivers consistently accurate results and expands analytical capability
AUSTIN, Texas, March 1, 2021 /PRNewswire/ -- Analytical testing laboratories performing high-throughput analysis in the food safety, environmental, pharmaceutical, anti-doping and forensic toxicology sectors can benefit from a simplified high resolution accurate mass gas chromatography-mass spectrometer (GC-MS) designed to provide sensitive data efficiently and confidently, allowing laboratories to speed-up turnaround time and expand their capabilities into new analytical avenues.
The Thermo Scientific Orbitrap Exploris GC-MS brings high-resolution analysis into everyday testing in a compact and easy-to-use instrument, delivering leading sensitivity and mass resolving power up to 60,000. Through an analytical dynamic range across six orders, the new system can provide accurate quantitation and detection of chemical components at trace and high concentrations, for targeted and non-targeted applications. The capability to acquire accurate mass data in full scan allows for multiple compound identification points, simplifying data acquisition, facilitating retrospective data analysis and accelerating instrument set-up for increased uptime and productivity.
"Contract testing laboratories must deliver high-quality results within ever-shortening turnaround times, while also complying with evolving regulations and addressing novel target analytes," said Fabrizio Moltoni, vice president and general manager, applied analytical technologies, chromatography and mass spectrometry, Thermo Fisher Scientific. "In order to meet and exceed our customers' needs, the Orbitrap Exploris GC-MS simplifies data acquisition and consolidates separate targeted methods into a single full scan system—opening new business opportunities to respond to changing testing requirements, and improving productivity and profitability through increased throughput."
David Haas, senior project manager, Eurofins GfA Lab Services GmbH said, "The addition of the Orbitrap Exploris GC-MS to our laboratory has allowed us to easily reach our target limits of quantification to ensure safety and compliance, and the selective full scan data enables us to work more efficiently, especially through data processing."
For high-throughput analysis, the Orbitrap Exploris GC-MS provides:
  • Streamlined instrument setup to initiate, tune and calibrate in under five minutes.
  • Easy-to-use operation software and pre-defined method templates to reduce training needs.
  • Data certainty and the opportunity to quickly increase scope with multiple points of identification.
  • Integrated informatics solutions with Thermo Scientific Chromeleon Chromatography Data System (CDS) or Thermo Scientific TraceFinder software to efficiently take data through to result.
  • Configurable system available with or without MS/MS capability and with flexible maximum resolving power, upgradable in the field for easier scaling.
  • Testing for a range of applications, such as quantitative pesticides, persistent organic pollutants, sports doping, environmental contaminants and nitrosamines.
The new system, along with the Thermo Scientific Orbitrap Exploris GC 240, extends the Thermo Scientific Orbitrap Exploris portfolio of high-resolution accurate mass systems, which is now comprised of the Thermo Scientific Orbitrap Exploris 480 mass spectrometer, launched in 2019, and the Thermo Scientific Orbitrap Exploris 240 and Thermo Scientific Orbitrap Exploris 120 mass spectrometers, launched in 2020.
To learn more, please visit www.thermofisher.com/orbitrapexplorisgc or register to attend "Your GC-MS Future Today: A Sponsored Virtual Event" on March 2, 2021.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with annual revenue exceeding $30 billion. Our Mission is to enable our customers to make the world healthier, cleaner and safer. Whether our customers are accelerating life sciences research, solving complex analytical challenges, improving patient diagnostics and therapies or increasing productivity in their laboratories, we are here to support them. Our global team of more than 80,000 colleagues delivers an unrivaled combination of innovative technologies, purchasing convenience and pharmaceutical services through our industry-leading brands, including Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific, Unity Lab Services and Patheon. For more information, please visit www.thermofisher.com.
Media Contact Information:
Laura Bright
Thermo Fisher Scientific
+1 562-335-8318
laura.bright@thermofisher.com  
Janice Foley
BioStrata
+1 617-823-5555
jfoley@biostratamarketing.com
SOURCE Thermo Fisher Scientific
(opens in a new window)