India Media Room
SAN JOSE, Calif., March 31, 2020 /PRNewswire/ -- Scientists no longer need to limit routine trace element analysis workflow productivity with the launch of a new series of inductively coupled plasma optical emission spectroscopy (ICP-OES) instruments for food and beverage, consumer safety, industrial, environmental and pharmaceutical laboratories.
The Thermo Scientific iCAP PRO Series ICP-OES platform is designed to provide a fast, sensitive range of trace element analysis solutions capable of capturing the complete spectrum of high matrix samples in a single run, improving workflow productivity and reducing analysis costs. From standby to start-up in just five minutes, the new instruments reduce gas consumption within a vertical dual-purged optical pathway interface that has lower installation requirements due to its standard wall socket and low extraction flow rate.
"The analysis of trace elements in high matrix samples is essential for laboratory teams across a number of sectors to assure the quality, consistency and safety of products and the environment we live in, but it traditionally presents acute analytical and resource challenges that limit routine deployment," said Adrian Holley, director, product marketing, trace element analysis, chromatography and mass spectrometry, Thermo Fisher Scientific. "The launch of the iCAP PRO Series ICP-OES platform helps laboratories bypass traditional sample preparation requirements and the need to undertake multiple measurements, providing a range of trace element analysis solutions that enhance workflow productivity and reduce cost-per-sample."
Mike Dudley, laboratory supervisor, fluid analysis laboratory, Carter Machinery Company, a producer of lubricants for engines and other mechanical purposes, said, "We have been running the iCAP PRO Series ICP-OES platform for several weeks and are very pleased. We have reduced our run time by 36%. Our numbers are looking great, and my team is delighted with its performance."
Laboratories using the iCAP PRO Series ICP-OES instruments will also benefit by:
- Reducing the number of measurements per run to obtain a complete spectrum through the combination of a single optical slit and a charge injection device detector, delivering increased analysis speeds and readout in a reduced time frame.
- Maximizing instrument uptime with fast start-up, making analysis possible after just five minutes.
- Minimal recalibration of the instrument as a result of highly stable optics and polychromator.
For more information of the iCAP PRO Series ICP-OES platform, please visit www.thermofisher.com/ICP-OES.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with annual revenue exceeding $25 billion. Our Mission is to enable our customers to make the world healthier, cleaner and safer. Whether our customers are accelerating life sciences research, solving complex analytical challenges, improving patient diagnostics and therapies or increasing productivity in their laboratories, we are here to support them. Our global team of more than 75,000 colleagues delivers an unrivaled combination of innovative technologies, purchasing convenience and pharmaceutical services through our industry-leading brands, including Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific, Unity Lab Services and Patheon. For more information, please visit www.thermofisher.com.
Media Contact Information:
Laura Bright
Thermo Fisher Scientific
+1 562-335-8318
laura.bright@thermofisher.com
Janice Foley
BioStrata
+1 617-823-5555
jfoley@biostratamarketing.com
SOURCE Thermo Fisher Scientific