India Media Room
PHILADELPHIA, Nov. 1, 2021 /PRNewswire/ -- ASMS 2021 – Thermo Fisher Scientific, the world leader in serving science, and MSAID, the leader in proteomics artificial intelligence, have collaborated to provide proteomics researchers with advanced mass spectrometry software that generates market-leading biological insight from acquired data by substantially increasing peptide identification and quantitation capabilities using artificial intelligence (AI) and deep learning.
Thermo Scientific Proteome Discoverer 3.0 software with CHIMERYS by MSAID leverages artificial intelligence (AI) to substantially enhance the identification rate and number of unique peptide identifications in proteomics data. CHIMERYS identifies a minimal set of peptides that can explain the acquired tandem mass spectrum, in comparison to existing methodologies, which typically assume all peaks in a tandem mass spectrum are derived from a single peptide. This innovative approach provides a 1.8-fold increase in the number of unique peptide identifications and a 1.5-fold increase in the number of protein identifications for typical proteomics data sets when compared to existing tools. In addition to improved protein coverage and quantitation capabilities, Proteome Discoverer 3.0 software paired with CHIMERYS also facilitates faster data acquisition for increased sample throughput.
Thermo Fisher Scientific and MSAID are showcasing their new software solution during the 69th American Society for Mass Spectrometry (ASMS) Conference on Mass Spectrometry and Allied Topics, being held October 31-November 4, in the Pennsylvania Convention Center, Philadelphia, Pennsylvania.
"Previous technologies have been unable to fully interpret data generated using mass spectrometry, as mass spectra can contain fragments from multiple co-isolated peptides that cannot be identified using current algorithms," said August Specht, vice president, research and development, chromatography and mass spectrometry, Thermo Fisher Scientific. "By using Proteome Discoverer 3.0 software and CHIMERYS, scientists can now leverage AI for deeper mining of proteomic data. This not only improves proteomic coverage, but also expands the ways in which proteomics scientists can acquire and apply their data."
Martin Frejno, chief executive officer, MSAID said: "Chimeric spectra were a long-standing problem in mass spectrometry-based proteomics. With CHIMERYS, we reimagined the analysis of tandem mass spectra from the ground up using AI to solve it."
The Proteome Discoverer 3.0 software release also includes an updated INFERYS prediction model, extending support to tandem mass tagging (TMT), collisionally induced dissociation (CID), and providing improved results for immunopeptidomics. By pairing intelligent data analysis through Proteome Discoverer 3.0 software and CHIMERYS with leading hardware technology in the Thermo Scientific Vanquish Neo Ultra-High Performance Liquid Chromatography (UHPLC) system and Thermo Scientific Orbitrap mass spectrometry platforms, researchers will be empowered to continue pushing the boundaries of proteomics research.
For more information on Thermo Scientific Proteome Discoverer 3.0 software and CHIMERYS by MSAID, please visit www.thermofisher.com/proteomediscoverer, or join us at ASMS 2021 from October 31-November 4, 2021.
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with annual revenue of approximately $35 billion. Our Mission is to enable our customers to make the world healthier, cleaner and safer. Whether our customers are accelerating life sciences research, solving complex analytical challenges, improving patient diagnostics and therapies or increasing productivity in their laboratories, we are here to support them. Our global team of more than 90,000 colleagues delivers an unrivaled combination of innovative technologies, purchasing convenience and pharmaceutical services through our industry-leading brands, including Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific, Unity Lab Services and Patheon. For more information, please visit www.thermofisher.com.
Media Contact Information:
Laura Bright
Thermo Fisher Scientific
+1 562-335-8318
laura.bright@thermofisher.com
Janice Foley
+1 617-823-5555
jfoley@biostratamarketing.com
SOURCE Thermo Fisher Scientific