India Media Room
VANTAA, Finland, March 16, 2020 /PRNewswire/ -- Thermo Fisher Scientific, the world leader in serving science, today announced that its Thermo Scientific Cascadion SM Clinical Analyzer is now commercially available in the U.S. with the Cascadion SM 25-Hydroxy Vitamin D assay. Designed to deliver accurate, traceable measurements of vitamin D2 (ergocalciferol) and D3 (cholecalciferol), this serum assay reports total vitamin D results alongside its components. Clinical laboratories can now benefit from a complete system bringing the accuracy, sensitivity and specificity benefits of Thermo Fisher's gold standard liquid chromatography-tandem mass spectrometry (LC-MS/MS) technology into routine clinical workflows.
Incorporating sample processing, LC separation and MS detection capabilities in a single solution, the Cascadion system is fully automated and enables load-up and walkaway sample management, eliminating the need for multiple connected components. The system offers end-to-end automation that is easy to use by any qualified laboratory personnel with minimal training, thus streamlining and optimizing the analytical process.
The Cascadion system analyzes samples directly from qualified primary blood collection tubes, while the Cascadion SM 25-Hydroxy Vitamin D Assay incorporates fully barcoded components, all traceable to a specific result. The vitamin D calibrators and controls are traceable according to the National Institute of Standards and Technology (NIST) and assay calibration remains stable for up to 30 days.
"The Centers for Disease Control and Prevention has established LC-MS/MS as the reference method for vitamin D testing," explains Bill Östman, vice president and general manager, Thermo Fisher Scientific. "However, LC-MS/MS has traditionally been a sophisticated technology, requiring specialized knowledge and significant hands-on time for method development. We developed the Cascadion system and the ready-to-use, pre-validated 25-Hydroxy Vitamin D assay to simplify and accelerate the implementation of LC-MS/MS in clinical settings without the need for extensive technical expertise."
The Cascadion analyzer is listed with the U.S. Food and Drug Administration (FDA) as a Class I instrument, while the Cascadion SM 25-Hydroxy Vitamin D assay is FDA-listed as Class II exempt with special controls.
For more information about the Cascadion SM Clinical Analyzer and Cascadion SM 25-Hydroxy Vitamin D Assay, please visit www.thermofisher.com/cascadion
About Thermo Fisher Scientific
Thermo Fisher Scientific Inc. is the world leader in serving science, with annual revenue exceeding $25 billion. Our Mission is to enable our customers to make the world healthier, cleaner and safer. Whether our customers are accelerating life sciences research, solving complex analytical challenges, improving patient diagnostics and therapies or increasing productivity in their laboratories, we are here to support them. Our global team of more than 75,000 colleagues delivers an unrivaled combination of innovative technologies, purchasing convenience and pharmaceutical services through our industry-leading brands, including Thermo Scientific, Applied Biosystems, Invitrogen, Fisher Scientific, Unity Lab Services and Patheon. For more information, please visit www.thermofisher.com.
Media Contact Information:
Julia Nisbet-Fahy
Senior Manager - Global Marketing
Thermo Fisher Scientific
+44 (0) 7342 080967
julia.nisbet-fahy@thermofisher.com
Jaime Grigorescu
Marketing Manager – Global Marketing
Thermo Fisher Scientific
+1 (773) 431 0390
jaime.grigorescu@thermofisher.com
SOURCE Thermo Fisher Scientific